Learn More
4-Hydroxyphenytoin (5-(4-Hydroxyphenyl)-5-phenylhydantoin), LoGiCal
Product Code. 30459301
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459301 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459301 Supplier LoGiCal Supplier No. LGCFOR0630.09
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-(4-Hydroxyphenyl)-5-phenylhydantoin
CAS 2784-27-2
Analyte or Component Names 5-(4-Hydroxyphenyl)-5-phenylhydantoin
Shipping Condition Room Temperature
Molecular Formula C15 H12 N2 O3
Certifications/Compliance ISO 9001
Synonym Hydantoin, 5-(p-hydroxyphenyl)-5-phenyl-, (p-Hydroxyphenyl)phenylhydantoin, (±)-5-(4-Hydroxyphenyl-5-phenylhydantoin, 4-Hydroxydiphenylhydantoin, 4-Hydroxyphenytoin, 5-(4-Hydroxyphenyl)-5-phenylhydantoin, 5-(4'-Hydroxyphenyl)-5-phenylhydantoin, 5-(p-Hydroxyphenyl)-5-phenylhydantoin, 5-Phenyl-5-p-hydroxyphenylhydantoin, Hydroxydiphenylhydantoin, NSC 156081, dl-5-(p-Hydroxyphenyl-5-phenylhydantoin, p-Hydroxydiphenylhydantoin, p-Hydroxyphenytoin, 2,4-Imidazolidinedione, 5-(4-hydroxyphenyl)-5-phenyl-
InChI Formula InChI=1S/C15H12N2O3/c18-12-8-6-11(7-9-12)15(10-4-2-1-3-5-10)13(19)16-14(20)17-15/h1-9,18H,(H2,16,17,19,20)
Solution Type Neat
SMILES Oc1ccc(cc1)C2(NC(=O)NC2=O)c3ccccc3
IUPAC Name 5-(4-hydroxyphenyl)-5-phenylimidazolidine-2,4-dione
Industry Type Clinical Testing
Molecular Weight (g/mol) 268.2674
Quantity 5 mg
Formula Weight 268.0848 g/mol