Learn More
4-Fluorothiobenzamide, 97%
Product Code. 15404777
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15404777
Brand: Thermo Scientific Alfa Aesar H32093.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 22179-72-2 | C7H6FNS | 155.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22179-72-2
Molecular Formula C7H6FNS
Molecular Weight (g/mol) 155.19
MDL Number MFCD01313285
InChI Key VQFOHZWOKJQOGO-UHFFFAOYSA-N
Synonym 4-fluorothiobenzamide, 4-fluorobenzothioamide, 4-fluorobenzene-1-carbothioamide, 4-fluoro-1-benzenecarbothioamide, 4-fluoro-thiobenzamide, benzenecarbothioamide, 4-fluoro, amino 4-fluorophenyl methane-1-thione, p-fluorothiobenzamide, acmc-20amhq, 4-fluoro thiobenzamide
PubChem CID 737223
IUPAC Name 4-fluorobenzenecarbothioamide
SMILES NC(=S)C1=CC=C(F)C=C1
Specifications
Melting Point 146°C to 148°C
Quantity 1 g
UN Number UN2811
Formula Weight 155.19
Percent Purity 97%
Chemical Name or Material 4-Fluorothiobenzamide