accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 22179-72-2

H2NSF

CAS RN 22179-72-2

IUPAC Name: 4-fluorobenzene-1-carbothioamide
Synonyms: 4-fluorobenzenecarbothioamide
Molecular Weight (g/mol): 155.19
Molecular Formula: C7H6FNS
InChi Key: VQFOHZWOKJQOGO-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results