missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 11338675
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338675 5 g 5g
11328675 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338675 Supplier Thermo Scientific Alfa Aesar Supplier No. L12214.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2740-88-7
Molecular Formula C8H6FNS
Molecular Weight (g/mol) 167.201
MDL Number MFCD00041115
InChI Key LPVNPJMEWWUFHD-UHFFFAOYSA-N
Synonym 4-fluorobenzyl isothiocyanate, 1-fluoro-4-isothiocyanatomethyl benzene, 4-fluorobenzylisothiocyanate, p-fluorobenzyl isothiocyanate, benzene, 1-fluoro-4-isothiocyanatomethyl, 4-fluorophenyl methanisothiocyanate, pubchem5031, acmc-20ami3, 4-fluorobenzyl-isothiocyanate, 4-isothiocyanatomethyl fluorobenzene
PubChem CID 75963
IUPAC Name 1-fluoro-4-(isothiocyanatomethyl)benzene
SMILES C1=CC(=CC=C1CN=C=S)F

Specifications

Density 1.22
Boiling Point 124°C to 125°C (6 mmHg)
Odor Irritating
Refractive Index 1.579
Quantity 5 g
UN Number UN2922
Beilstein 2085915
Sensitivity Moisture sensitive
Formula Weight 167.2
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.