accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FNCS

CAS RN 2740-88-7

IUPAC Name: 1-fluoro-4-(isothiocyanatomethyl)benzene
Synonyms: 1-fluoro-4-(isothiocyanatomethyl)benzene
Molecular Weight (g/mol): 167.2
Molecular Formula: C8H6FNS
InChi Key: LPVNPJMEWWUFHD-UHFFFAOYSA-N
SMILES: FC1=CC=C(CN=C=S)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results