missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-methylphenyl)benzamide, 97%, Thermo Scientific™

Product Code. 15493718
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493718 250 mg 250mg
15483718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493718 Supplier Thermo Scientific Alfa Aesar Supplier No. H57191.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-05-3
Molecular Formula C14H12FNO
Molecular Weight (g/mol) 229.254
MDL Number MFCD00446355
InChI Key FYIMLKPLYFXRDM-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-methylphenyl benzamide, 4-fluoro-n-p-tolyl-benzamide, 4-fluoro-n-p-tolyl benzamide, n-4-methylphenyl-4-fluorobenzamide, benzamide,4-fluoro-n-4-methylphenyl, 4-fluoro-n∼1∼-4-methylphenyl benzamide, 4-fluorophenyl-n-4-methylphenyl carboxamide
PubChem CID 693096
IUPAC Name 4-fluoro-N-(4-methylphenyl)benzamide
SMILES CC1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)F

Specifications

Quantity 250 mg
Formula Weight 229.25
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-(4-methylphenyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.