accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNOF

CAS RN 399-05-3

IUPAC Name: 4-fluoro-N-(4-methylphenyl)benzamide
Synonyms: 4-fluoro-N-(4-methylphenyl)benzamide
Molecular Weight (g/mol): 229.25
Molecular Formula: C14H12FNO
InChi Key: FYIMLKPLYFXRDM-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(=O)C2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results