missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™

Product Code. 15408615
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15408615 1 g 1g
15418615 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15408615

Brand: Thermo Scientific Alfa Aesar H33725.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373921-81-3
Molecular Formula C11H8F4O2
Molecular Weight (g/mol) 248.177
MDL Number MFCD22201075
InChI Key JWTWWSLPXHUYNI-NSCUHMNNSA-N
Synonym 4-fluoro-3-methyl-5-trifluoromethyl cinnamic acid, 2e-3-4-fluoro-3-methyl-5-trifluoromethyl phenyl prop-2-enoic acid, 3-4-fluoro-3-methyl-5-trifluoromethyl phenyl acrylic acid
PubChem CID 86277682
IUPAC Name (E)-3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES CC1=C(C(=CC(=C1)C=CC(=O)O)C(F)(F)F)F

Specifications

Quantity 1 g
Formula Weight 248.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.