CAS RN 1373921-81-3
CAS RN 1373921-81-3
IUPAC Name:
3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms:
3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol):
248.18
Molecular Formula:
C11H8F4O2
InChi Key:
JWTWWSLPXHUYNI-UHFFFAOYSA-N
SMILES:
CC1=C(F)C(=CC(C=CC(O)=O)=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™
CAS: 1373921-81-3 Molecular Formula: C11H8F4O2 Molecular Weight (g/mol): 248.177 MDL Number: MFCD22201075 InChI Key: JWTWWSLPXHUYNI-NSCUHMNNSA-N Synonym: 4-fluoro-3-methyl-5-trifluoromethyl cinnamic acid,2e-3-4-fluoro-3-methyl-5-trifluoromethyl phenyl prop-2-enoic acid,3-4-fluoro-3-methyl-5-trifluoromethyl phenyl acrylic acid PubChem CID: 86277682 IUPAC Name: (E)-3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid SMILES: CC1=C(C(=CC(=C1)C=CC(=O)O)C(F)(F)F)F