Learn More
4-Fluoro-3,5-dimethylbenzaldehyde, 97%, Thermo Scientific™
Product Code. 11346979
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346979 5 g 5g
11336979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11346979 Supplier Thermo Scientific Alfa Aesar Supplier No. H26832.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 363134-35-4
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD06660264
InChI Key NNHYTIOMCJAWLM-UHFFFAOYSA-N
Synonym benzaldehyde, 4-fluoro-3,5-dimethyl, acmc-1ctao, 3,5-dimethyl-4-fluorobenzaldehyde, 4-fluoro-3,5-dimethyl-benzaldehyde, benzaldehyde, 4-fluoro-3,5-dimethyl-9ci
PubChem CID 12040710
IUPAC Name 4-fluoro-3,5-dimethylbenzaldehyde
SMILES CC1=CC(=CC(=C1F)C)C=O
Specifications
Refractive Index 1.523
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3,5-dimethylbenzaldehyde