accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3FH3CO

CAS RN 363134-35-4

IUPAC Name: 4-fluoro-3,5-dimethylbenzaldehyde
Synonyms: 4-fluoro-3,5-dimethylbenzaldehyde
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: NNHYTIOMCJAWLM-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=CC(C)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results