Learn More
4-Desmethyl-4-ethylimidazolyl Nilotinib, TRC
Product Code. 17964395
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964395 100 mg 100mg
17950405 25 mg 25mg
17993915 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17964395 Supplier Toronto Research Chemicals Supplier No. TRCD291210100MG
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2119583-24-1
Molecular Formula C29H24F3N7O
Molecular Weight (g/mol) 543.54
Synonym Benzamide, N-[3-(4-ethyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, N-[3-(4-Ethyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzamide, Nilotinib ethyl homolog
SMILES O=C(NC1=CC(C(F)(F)F)=CC(N2C=C(CC)N=C2)=C1)C3=CC=C(C)C(NC4=NC(C5=CN=CC=C5)=CC=N4)=C3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C29H24F3N7O/c1-3-22-16-39(17-35-22)24-13-21(29(30,31)32)12-23(14-24)36-27(40)19-7-6-18(2)26(11-19)38-28-34-10-8-25(37-28)20-5-4-9-33-15-20/h4-17H,3H2,1-2H3,(H,36,40)(H,34,37,38)
Chemical Name or Material 4-Desmethyl-4-ethylimidazolyl Nilotinib