CAS RN 2119583-24-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Desmethyl-4-ethylimidazolyl Nilotinib, TRC
CAS: 2119583-24-1 Molecular Formula: C29H24F3N7O Molecular Weight (g/mol): 543.54 Synonym: Benzamide, N-[3-(4-ethyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-,N-[3-(4-Ethyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzamide,Nilotinib ethyl homolog SMILES: O=C(NC1=CC(C(F)(F)F)=CC(N2C=C(CC)N=C2)=C1)C3=CC=C(C)C(NC4=NC(C5=CN=CC=C5)=CC=N4)=C3