missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyclopropylsulfamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15419937
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419937 1 g 1g
15400446 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419937 Supplier Thermo Scientific Alfa Aesar Supplier No. H53170.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871329-67-8
Molecular Formula C9H12BNO4S
Molecular Weight (g/mol) 241.07
MDL Number MFCD07363755
InChI Key FUEGEHDCXRTJPN-UHFFFAOYSA-N
Synonym n-cyclopropyl 4-boronobenzenesulfonamide, 4-cyclopropylsulfamoyl phenylboronic acid, 4-n-cyclopropylsulfamoyl phenyl boronic acid, 4-n-cyclopropylsulfamoyl phenylboronic acid, 4-cyclopropylsulfamoyl phenyl boronic acid, 4-borono-n-cyclopropylbenzenesulphonamide, 4-n-cyclopropylsulphonamido benzeneboronic acid, boronic acid,b-4-cyclopropylamino sulfonyl phenyl, acmc-209qgb, 4-n-cyclopropylsulfamoyl phenyl boronicacid
PubChem CID 44119553
IUPAC Name [4-(cyclopropylsulfamoyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)S(=O)(=O)NC1CC1

Specifications

Melting Point 142°C to 144°C
Quantity 1 g
Formula Weight 241.07
Percent Purity 98%
Chemical Name or Material 4-(Cyclopropylsulfamoyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.