missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenol, TRC

Product Code. 17939629
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939629 1 g 1g
30136191 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17939629 Supplier Toronto Research Chemicals Supplier No. TRCC3750401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-48-9
Formula Weight 128.0029
InChI Formula InChI=1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H
IUPAC Name 4-chlorophenol
Molecular Formula C6 H5 Cl O
Molecular Weight (g/mol) 128.56
SMILES Oc1ccc(Cl)cc1
Synonym Phenol, p-chloro- (8CI), 4-Chlorophenol, 4-CP, 4-Hydroxychlorobenzene, Applied 3-78, NSC 2877, p-Chlorophenic acid, p-Chlorophenol
Chemical Name or Material 4-Chlorophenol

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.