missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic Acid, TRC

Product Code. 17923585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923585 1 g 1g
30135435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17923585 Supplier Toronto Research Chemicals Supplier No. TRCC3827001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1205-30-7
Formula Weight 234.9706
InChI Formula InChI=1S/C7H6ClNO4S/c8-5-2-1-4(7(10)11)3-6(5)14(9,12)13/h1-3H,(H,10,11)(H2,9,12,13)
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
Molecular Formula C7 H6 Cl N O4 S
Molecular Weight (g/mol) 235.64
Percent Purity >95%
SMILES NS(=O)(=O)c1cc(ccc1Cl)C(=O)O
Synonym 4-Chloro-3-sulfamoylbenzoic Acid, Benzoic acid, 3-(aminosulfonyl)-4-chloro- (9CI, ACI), 3-(Aminosulfonyl)-4-chlorobenzoic acid (ACI), Benzoic acid, 4-chloro-3-sulfamoyl- (7CI, 8CI), 3-Sulfamoyl-4-chlorobenzoic acid, 3-Sulfamyl-4-chlorobenzoic acid, 4-Chloro-3-(aminosulfonyl)benzoic acid, 4-Chloro-3-sulfamoylbenzoic acid, 4-Chloro-5-sulfamoylbenzoic acid
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic Acid
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.