missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonamide, Maybridge

Product Code. 10137314
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10137314 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10137314 Supplier Thermo Scientific Maybridge Supplier No. MAY00130DE
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 701-34-8
Packaging Amber Glass Bottle
Molecular Formula C6H6BrNO2S
Quantity 5 g
Synonym p-bromobenzenesulfonamide, benzenesulfonamide, p-bromo, benzenesulfonamide, 4-bromo, 4-bromobenzene-1-sulfonamide, 4-bromo-benzenesulfonamide, 4-bromobenzene sulfonamide, pbsa, 4-bromophenylsulfonamide, 4-bromo-phenylsulfonamide, wln: zswr de
InChI Key STYQHICBPYRHQK-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1S(=O)(=O)N)Br
IUPAC Name 4-bromobenzenesulfonamide
Molecular Weight (g/mol) 236.083
PubChem CID 69696
Formula Weight 236.08
Percent Purity 97%
Chemical Name or Material 4-Bromobenzenesulfonamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.