missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)anisole, 97%, Thermo Scientific™

Product Code. 11475887
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475887 10 g 10g
11465887 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475887 Supplier Thermo Scientific Alfa Aesar Supplier No. B24436.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1514-11-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD01631579
InChI Key XTFPWZDSCKENBJ-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl anisole, 4-bromo-1-methoxy-2-trifluoromethyl benzene, 4-methoxy-3-trifluoromethyl bromobenzene, 5-bromo-2-methoxybenzotrifluoride, 4-bromo-1-methoxy-2-trifluoromethyl-benzene, 2-trifluoromethyl-4-bromoanisole, 3-bromo-6-methoxybenzotrifluoride, 1-bromo-4-methoxy-3-trifluoromethyl benzene, acmc-20amfg, pubchem4441
PubChem CID 2775304
IUPAC Name 4-bromo-1-methoxy-2-(trifluoromethyl)benzene
SMILES COC1=CC=C(Br)C=C1C(F)(F)F

Specifications

Boiling Point 120°C (30 mmHg)
Refractive Index 1.498
Quantity 10 g
Beilstein 2525206
Formula Weight 255.04
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)anisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.