accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OBrFFF

CAS RN 1514-11-0

IUPAC Name: 4-bromo-1-methoxy-2-(trifluoromethyl)benzene
Synonyms: 4-bromo-1-methoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: XTFPWZDSCKENBJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results