missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4'-(Boc-amino)biphenyl-3-carboxylic acid, 95%

Product Code. p-7034250
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438407 1 g 1g
15448407 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438407 Supplier Alfa Aesar™ Supplier No. H51918.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 927801-51-2
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.353
MDL Number MFCD09055096
InChI Key ZBRGFWLFMFHGGR-UHFFFAOYSA-N
Synonym 3-4-tert-butoxycarbonyl amino phenyl benzoic acid, 4'-tert-butoxycarbonyl amino-1,1'-biphenyl-3-carboxylic acid, 4'-boc-amino biphenyl-3-carboxylic acid, 4 boc-amino biphenyl-3-carboxylic acid, 4'-tert-butoxycarbonyl amino-3-carboxybiphenyl, 4'-tert-butoxycarbonyl amino biphenyl-3-carboxylic acid, 4'-tert-butoxycarbonyl amino 1,1'-biphenyl-3-carboxylic acid
PubChem CID 53218487
IUPAC Name 3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 313.35
Percent Purity 95%
Chemical Name or Material 4'-(Boc-amino)biphenyl-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.