accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 927801-51-2

H3CH3CCH3OONHOOH

CAS RN 927801-51-2

IUPAC Name: 4'-{[(tert-butoxy)carbonyl]amino}-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 4'-[(tert-butoxycarbonyl)amino]-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 313.35
Molecular Formula: C18H19NO4
InChi Key: ZBRGFWLFMFHGGR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(C=C1)C1=CC(=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results