Learn More
4-Benzyloxy-2-hydroxybenzaldehyde, 99%
Product Code. 11368296
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368296 1 g 1g
11378296 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11368296 Supplier Thermo Scientific Alfa Aesar Supplier No. L15515.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 52085-14-0 | C14H12O3 | 228.247 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52085-14-0
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD01075698
InChI Key AMLKEDBYDOCGEG-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-hydroxybenzaldehyde, benzaldehyde, 2-hydroxy-4-phenylmethoxy, 4-benzyloxy-2-hydroxy-benzaldehyde, 2-hydroxy-4-phenylmethoxy benzaldehyde, acmc-20amov, 4-benzyloxysalicylaldehyde, 4-benzyloxy-2-hydoxy-benzaldehyde, 4-bezyloxy-2-hydroxy-benzaldehyde, 4-benzyloxy-2-hydroxy benzaldehyde
PubChem CID 561235
IUPAC Name 2-hydroxy-4-phenylmethoxybenzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC(=C(C=C2)C=O)O
Specifications
Melting Point 77°C to 80°C
Quantity 1 g
Beilstein 982995
Sensitivity Air Sensitive
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material 4-Benzyloxy-2-hydroxybenzaldehyde