accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOHO

CAS RN 52085-14-0

IUPAC Name: 4-(benzyloxy)-2-hydroxybenzaldehyde
Synonyms: 4-(benzyloxy)-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: AMLKEDBYDOCGEG-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=CC(OCC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results