Learn More
4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester, 95%, Thermo Scientific™
Product Code. 15457675
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15457675
Brand: Thermo Scientific Alfa Aesar H27292.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1073354-22-9
Molecular Formula C17H20BClN2O3
Molecular Weight (g/mol) 346.618
MDL Number MFCD06798267
InChI Key YXXPVSCUVOPQBT-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 4-benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester, 4-benzyloxy-2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloro-4-phenoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, abbypharma ap-31-6586, 2-chloro-4-phenylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine
PubChem CID 44755193
IUPAC Name 2-chloro-4-phenylmethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2OCC3=CC=CC=C3)Cl
Specifications
Melting Point 99°C to 101°C
Quantity 250 mg
Formula Weight 346.62
Percent Purity 95%
Chemical Name or Material 4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester