accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1073354-22-9

BOOH3CCH3H3CH3CNNOCl

CAS RN 1073354-22-9

IUPAC Name: 4-(benzyloxy)-2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Synonyms: 4-(benzyloxy)-2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Molecular Weight (g/mol): 346.62
Molecular Formula: C17H20BClN2O3
InChi Key: YXXPVSCUVOPQBT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results