missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyl(methyl)amino)benzoic Acid, TRC

Product Code. 30338447
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338447 100 mg 100mg
30343758 25 mg 25mg
30329379 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30338447 Supplier Toronto Research Chemicals Supplier No. TRCB287865100MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyl(methyl)amino)benzoic Acid
Analyte or Component Names 4-(Benzyl(methyl)amino)benzoic Acid
Shipping Condition Room Temperature
Molecular Formula C15 H15 N O2
InChI Formula InChI=1S/C15H15NO2/c1-16(11-12-5-3-2-4-6-12)14-9-7-13(8-10-14)15(17)18/h2-10H,11H2,1H3,(H,17,18)
Solution Type Neat
SMILES CN(Cc1ccccc1)c2ccc(cc2)C(=O)O
IUPAC Name 4-[benzyl(methyl)amino]benzoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 241.285
Quantity 100 mg
Formula Weight 241.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.