Learn More
4-Amino-5,6-dichlorobenzene-1,3-disulfonamide, Mikromol™
Description
4-Amino-5,6-dichlorobenzene-1,3-disulfonamide
Specifications
Specifications
| Chemical Name or Material | 4-Amino-5,6-dichlorobenzene-1,3-disulfonamide |
| CAS | 5250-72-6 |
| Analyte or Component Names | 4-Amino-5,6-dichlorobenzene-1,3-disulfonamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H7 Cl2 N3 O4 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,3-Benzenedisulfonamide, 4-amino-5,6-dichloro-, m-Benzenedisulfonamide, 4-amino-5,6-dichloro- (6CI,7CI,8CI), 4-Amino-5,6-dichloro-1,3-benzenedisulfonamide, 4-Amino-5,6-dichlorobenzene-1,3-disulfonamide |
| InChI Formula | InChI=1S/C6H7Cl2N3O4S2/c7-4-2(16(10,12)13)1-3(17(11,14)15)6(9)5(4)8/h1H,9H2,(H2,10,12,13)(H2,11,14,15) |
| SMILES | Nc1c(Cl)c(Cl)c(cc1S(=O)(=O)N)S(=O)(=O)N |
| IUPAC Name | 4-amino-5,6-dichlorobenzene-1,3-disulfonamide |
| Show More |