Learn More
4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride, Mikromol™
Product Code. 30462445
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462445 4 x 25 mg -
17924326 100 mg 100mg
17943006 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30462445 Supplier Mikromol Supplier No. MM0329.0400254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride
CAS 138564-60-0
Analyte or Component Names 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H11 N3 S . Cl H
Certifications/Compliance ISO 17025
Synonym 10H-Thieno[2,3-b][1,5]benzodiazepin-4-amine, 2-methyl-, hydrochloride (1:1), 10H-Thieno[2,3-b][1,5]benzodiazepin-4-amine, 2-methyl-, monohydrochloride (9CI), 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine hydrochloride, 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine monohydrochloride, Olanzamine
InChI Formula InChI=1S/C12H11N3S.ClH/c1-7-6-8-11(13)14-9-4-2-3-5-10(9)15-12(8)16-7;/h2-6,15H,1H3,(H2,13,14);1H
Solution Type Neat
SMILES Cl.Cc1cc2C(=Nc3ccccc3Nc2s1)N
IUPAC Name 2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-amine;hydrochloride
Molecular Weight (g/mol) 265.76
Quantity 4 x 25 mg
Formula Weight 265.044 g/mol