Learn More
4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride, Mikromol™
Description
4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride |
| CAS | 138564-60-0 |
| Analyte or Component Names | 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H11 N3 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 10H-Thieno[2,3-b][1,5]benzodiazepin-4-amine, 2-methyl-, hydrochloride (1:1), 10H-Thieno[2,3-b][1,5]benzodiazepin-4-amine, 2-methyl-, monohydrochloride (9CI), 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine hydrochloride, 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine monohydrochloride, Olanzamine |
| InChI Formula | InChI=1S/C12H11N3S.ClH/c1-7-6-8-11(13)14-9-4-2-3-5-10(9)15-12(8)16-7;/h2-6,15H,1H3,(H2,13,14);1H |
| SMILES | Cl.Cc1cc2C(=Nc3ccccc3Nc2s1)N |
| IUPAC Name | 2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-amine;hydrochloride |
| Show More |