Learn More
4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazapine, Hydrochloride, TRC
Product Code. 17992825
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992825 100 mg 100mg
17941735 250 mg 250mg
17963825 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17992825 Supplier Toronto Research Chemicals Supplier No. TRCA618255100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138564-60-0
Molecular Formula C12 H11 N3 S . Cl H
Molecular Weight (g/mol) 265.76
Synonym 10H-Thieno[2,3-b][1,5]benzodiazepin-4-amine, 2-methyl-, hydrochloride (1:1), 10H-Thieno[2,3-b][1,5]benzodiazepin-4-amine, 2-methyl-, monohydrochloride (9CI), 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine hydrochloride, 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine monohydrochloride, Olanzamine
IUPAC Name 2-methyl-10H-thieno[2,3-b][1,5]benzodiazepin-4-amine;hydrochloride
SMILES Cl.Cc1cc2C(=Nc3ccccc3Nc2s1)N
Specifications
Analyte or Component Names 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C12H11N3S.ClH/c1-7-6-8-11(13)14-9-4-2-3-5-10(9)15-12(8)16-7;/h2-6,15H,1H3,(H2,13,14);1H
Solution Type Neat
Industry Type Forensics
Quantity 100 mg
Formula Weight 265.044
Chemical Name or Material 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]-benzodiazepine Hydrochloride