Learn More
4-Amino-2-bromopyridine, 97%
Product Code. 11314396
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314396 1 g 1g
11324396 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11314396 Supplier Thermo Scientific Alfa Aesar Supplier No. L20007.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 7598-35-8 | C5H5BrN2 | 173.01 g/mol
4-Amino-2-bromopyridine is used as medical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-2-bromopyridine is used as medical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7598-35-8
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD01646061
InChI Key GNTGEMWEXKBWBX-UHFFFAOYSA-N
Synonym 4-amino-2-bromopyridine, 2-bromo-4-aminopyridine, 2-bromo-4-pyridylamine, 2-bromo-4-pyridinamine, 4-pyridinamine, 2-bromo, 2-bromo-pyridin-4-ylamine, pubchem1100, pubchem1222, 2-brompyridin-4-amin, 2-bromopyridine-4-amine
PubChem CID 346455
IUPAC Name 2-bromopyridin-4-amine
SMILES NC1=CC=NC(Br)=C1
Specifications
Melting Point 96°C to 99°C
Quantity 1 g
Beilstein 108928
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 97%
Chemical Name or Material 4-Amino-2-bromopyridine