accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7598-35-8

IUPAC Name: 2-bromopyridin-4-amine
Synonyms: 2-bromopyridin-4-amine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: GNTGEMWEXKBWBX-UHFFFAOYSA-N
SMILES: NC1=CC=NC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results