Learn More
4-(7,9-Dioxo-8-azaspiro[4.5]dec-8-yl)butyl [1-[2-Oxo-2-[[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl]amino]ethyl]cyclopentyl]acetate Oxalate, Mikromol™
Description
4-(7,9-Dioxo-8-azaspiro[4.5]dec-8-yl)butyl [1-[2-Oxo-2-[[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl]amino]ethyl]cyclopentyl]acetate Oxalate
Specifications
Specifications
| Chemical Name or Material | 4-(7,9-Dioxo-8-azaspiro[4.5]dec-8-yl)butyl [1-[2-Oxo-2-[[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl]amino]ethyl]cyclopentyl]acetate Oxalate |
| CAS | 2726492-73-3 |
| Analyte or Component Names | 4-(7,9-Dioxo-8-azaspiro[4.5]dec-8-yl)butyl [1-[2-Oxo-2-[[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl]amino]ethyl]cyclopentyl]acetate Oxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H52 N6 O5 . C2 H2 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Buspirone Imp. J (EP) as Oxalate, 4-(7,9-Dioxo-8-azaspiro[4.5]dec-8-yl)butyl [1-[2-Oxo-2-[[4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl]-amino]ethyl]cyclopentyl]acetate Oxalate |
| InChI Formula | InChI=1S/C34H52N6O5.C2H2O4/c41-28(35-14-5-6-17-38-19-21-39(22-20-38)32-36-15-9-16-37-32)24-33(10-1-2-11-33)27-31(44)45-23-8-7-18-40-29(42)25-34(26-30(40)43)12-3-4-13-34;3-1(4)2(5)6/h9,15-16H,1-8,10-14,17-27H2,(H,35,41);(H,3,4)(H,5,6) |
| SMILES | OC(=O)C(=O)O.O=C(CC1(CC(=O)OCCCCN2C(=O)CC3(CCCC3)CC2=O)CCCC1)NCCCCN4CCN(CC4)c5ncccn5 |
| IUPAC Name | 4-(7,9-dioxo-8-azaspiro[4.5]decan-8-yl)butyl 2-[1-[2-oxo-2-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butylamino]ethyl]cyclopentyl]acetate;oxalic acid |
| Show More |