Learn More
4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30322444
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322444 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30322444 Supplier Dr. Ehrenstorfer Supplier No. DREA14233020ME2000
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Lyral (mixture of isomers)
CAS 31906-04-4
Analyte or Component Names Lyral (mixture of isomers)
Shipping Condition Room Temperature
Molecular Formula C13 H22 O2
Concentration 2000 μg/mL
Certifications/Compliance ISO 17034
Synonym 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexenecarboxaldehyde, 4-(4-Methyl-4-hydroxyamyl)cyclohex-3-ene carboxaldehyde, Landolal, Liral
InChI Formula InChI=1S/C13H22O2/c1-13(2,15)9-3-4-11-5-7-12(10-14)8-6-11/h5,10,12,15H,3-4,6-9H2,1-2H3
SMILES CC(C)(O)CCCC1=CCC(CC1)C=O
IUPAC Name 4-(4-hydroxy-4-methylpentyl)cyclohex-3-ene-1-carbaldehyde
Solvent or Matrix Methanol
Molecular Weight (g/mol) 210.31
Quantity 1 mL
Formula Weight 210.162 g/mol