Learn More
4-(2-Pyridyl)benzaldehyde, 98%
Product Code. 15421778
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421778 1 g 1g
15431778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15421778 Supplier Thermo Scientific Alfa Aesar Supplier No. H55498.03
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 127406-56-8 | C12H9NO | 183.21 g/mol
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir
Solubility
Soluble in Chloroform
Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 127406-56-8
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD01863537
InChI Key NMLYGLCBSFKJFI-UHFFFAOYSA-N
Synonym 4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde
PubChem CID 1515240
IUPAC Name 4-pyridin-2-ylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=N1
Specifications
Melting Point 50°C to 52°C
Flash Point 113°C (235°F)
Quantity 1 g
Solubility Information Soluble in Chloroform
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material 4-(2-Pyridyl)benzaldehyde