accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ON

CAS RN 127406-56-8

IUPAC Name: 4-(pyridin-2-yl)benzaldehyde
Synonyms: 4-(pyridin-2-yl)benzaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: NMLYGLCBSFKJFI-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results