Learn More
4-(2-Propoxybenzoylamino)-1-methyl-3-propyl-1H-pyrazole-5-carboxamide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4-(2-Propoxybenzoylamino)-1-methyl-3-propyl-1H-pyrazole-5-carboxamide
Specifications
Specifications
| Chemical Name or Material | 3-(((5-(Aminocarbonyl)-1-methyl-3-propyl-1H-pyrazol-4-yl)amino)carbonyl)-4-propoxybenzene |
| CAS | 139756-04-0 |
| Analyte or Component Names | 3-(((5-(Aminocarbonyl)-1-methyl-3-propyl-1H-pyrazol-4-yl)amino)carbonyl)-4-propoxybenzene |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18H24N4O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Methyl-4-(2-propoxybenzamido)-3-propylpyrazole-5-carboxamide, 1-Methyl-4-[(2-propoxybenzoyl)amino]-3-propyl-1H-pyrazole-5-carboxamide, 1H-Pyrazole-5-carboxamide, 1-methyl-4-[(2-propoxybenzoyl)amino]-3-propyl-, 1-Methyl-4-(2-propoxybenzamido)-3-propylpyrazole-5-carboxamide, 2-Methyl-4-(2-propoxybenzoylamino)-5-propyl-2H-pyrazole-3-carboxamide, 2-Methyl-4-[(2-propoxybenzoyl)amino]-5-propylpyrazole-3-carboxamide, 4-(2-Propoxybenzoylamino)-1-methyl-3-propyl-1H-pyrazole-5-carboxamide |
| InChI Formula | InChI=1S/C18H24N4O3/c1-4-8-13-15(16(17(19)23)22(3)21-13)20-18(24)12-9-6-7-10-14(12)25-11-5-2/h6-7,9-10H,4-5,8,11H2,1-3H3,(H2,19,23)(H,20,24) |
| SMILES | CCCOc1ccccc1C(=O)Nc2c(CCC)nn(C)c2C(=O)N |
| IUPAC Name | 2-methyl-4-[(2-propoxybenzoyl)amino]-5-propylpyrazole-3-carboxamide |
| Show More |