Learn More
4-[2-[2-Hydroxy-3-(isopropylamino)propoxy]ethyl]phenol, Mikromol™
Product Code. 17972866
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972866 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17972866 Supplier Mikromol Supplier No. MM0027.270025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[2-[2-Hydroxy-3-(isopropylamino)propoxy]ethyl]phenol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[2-[2-Hydroxy-3-(isopropylamino)-propoxy]ethyl]phenol
CAS 2732276-87-6
Analyte or Component Names 4-[2-[2-Hydroxy-3-(isopropylamino)-propoxy]ethyl]phenol
Shipping Condition Room Temperature
Molecular Formula C14 H23 N O3
Certifications/Compliance ISO 17025
Synonym 4-[2-[2-Hydroxy-3-(isopropylamino)propoxy]ethyl]phenol
InChI Formula InChI=1S/C14H23NO3/c1-11(2)15-9-14(17)10-18-8-7-12-3-5-13(16)6-4-12/h3-6,11,14-17H,7-10H2,1-2H3
SMILES CC(C)NCC(O)COCCc1ccc(O)cc1
IUPAC Name 4-[2-[2-hydroxy-3-(propan-2-ylamino)propoxy]ethyl]phenol
Molecular Weight (g/mol) 253.34
Quantity 25 mg
Formula Weight 253.1678 g/mol