Learn More
4-[(1RS)-2-(tert-Butylamino)-1-hydroxyethyl]-2-ethylphenol Hydrobromide, Mikromol™
Product Code. 17981486
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981486 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17981486 Supplier Mikromol Supplier No. MM0381.170025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[(1RS)-2-(tert-Butylamino)-1-hydroxyethyl]-2-ethylphenol Hydrobromide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[(1RS)-2-(tert-Butylamino)-1-hydroxyethyl]-2-ethylphenol Hydrobromide
CAS 2731709-15-0
Analyte or Component Names 4-[(1RS)-2-(tert-Butylamino)-1-hydroxyethyl]-2-ethylphenol Hydrobromide
Shipping Condition Room Temperature
Molecular Formula C14 H23 N O2 . Br H
Certifications/Compliance ISO 9001
Synonym 4-[(1RS)-2-(tert-Butylamino)-1-hydroxyethyl]-2-ethylphenol Hydrobromide
InChI Formula InChI=1S/C14H23NO2.BrH/c1-5-10-8-11(6-7-12(10)16)13(17)9-15-14(2,3)4;/h6-8,13,15-17H,5,9H2,1-4H3;1H
SMILES Br.CCc1cc(ccc1O)C(O)CNC(C)(C)C
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-ethylphenol;hydrobromide
Molecular Weight (g/mol) 318.25
Quantity 25 mg
Formula Weight 317.099 g/mol