Learn More
4-[(1RS)-1,3-Dimethylbutyl]-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one, Mikromol™
Description
4-[(1RS)-1,3-Dimethylbutyl]-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one
Specifications
Specifications
| Chemical Name or Material | 4-[(1RS)-1,3-Dimethylbutyl]-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one |
| CAS | 2731375-50-9 |
| Analyte or Component Names | 4-[(1RS)-1,3-Dimethylbutyl]-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H24 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Propyphenazone Imp. C (EP), 4-[(1RS)-1,3-Dimethylbutyl]-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one, Propyphenazone Impurity C |
| InChI Formula | InChI=1S/C17H22N2O.C4H6O4/c1-17(20-14-13-19(2)3,15-7-5-4-6-8-15)16-9-11-18-12-10-16;5-3(6)1-2-4(7)8/h4-12H,13-14H2,1-3H3;1-2H2,(H,5,6)(H,7,8) |
| SMILES | CC(C)CC(C)C1=C(C)N(C)N(C1=O)c2ccccc2 |
| IUPAC Name | 1,5-dimethyl-4-(4-methylpentan-2-yl)-2-phenylpyrazol-3-one |
| Show More |