missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[(3S)-3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride, Mikromol™
Description
[(3S)-3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride
Specifications
Specifications
| Chemical Name or Material | ((3S)-3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl)acetic Acid Hydrochloride |
| Analyte or Component Names | ((3S)-3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl)acetic Acid Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H34 N2 O5 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | [(3S)-3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride, 3-Amino-2,3,4,5-tetrahydro-2-oxo-1H-1-(3S)-benzazepine-1-acetic Acid Monohydrochloride, Benazepril Related Compound E (USP), Benazepril Hydrochloride Imp. E (EP), Benazepril Imp. E (EP), Benazepril USP Related Compound E, Benazepril USP RC E, [(3S)-3-[[(1S)-3-Cyclohexyl-1-(ethoxycarbonyl)propyl]amino]-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic Acid Hydrochloride, 3-(1-Ethoxycarbonyl-3-cyclohexyl-(1S)-propyl)amino-2,3,4,5-tetrahydro-2-oxo-1H-1-(3S)-benzazepine-1-acetic acid monohydrochloride, Benazepril Related Compound D (USP), Benazepril Related Compound E as Hydrochloride, Benazepril Hydrochloride Impurity E as Hydrochloride, Benazepril Impurity E as Hydrochloride |
| InChI Formula | InChI=1S/C24H34N2O5.ClH/c1-2-31-24(30)20(14-12-17-8-4-3-5-9-17)25-19-15-13-18-10-6-7-11-21(18)26(23(19)29)16-22(27)28;/h6-7,10-11,17,19-20,25H,2-5,8-9,12-16H2,1H3,(H,27,28);1H/t19-,20-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@H](CCC1CCCCC1)N[C@H]2CCc3ccccc3N(CC(=O)O)C2=O |
| IUPAC Name | 2-[(3S)-3-[[(2S)-4-cyclohexyl-1-ethoxy-1-oxobutan-2-yl]amino]-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 467 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?