Learn More
(3RS)-N,N-Bis(1-methylethyl)-3-phenyl-3-(pyridin-2-yl)propan-1-amine Dihydrogen Phosphate, Mikromol™
Description
(3RS)-N,N-Bis(1-methylethyl)-3-phenyl-3-(pyridin-2-yl)propan-1-amine Dihydrogen Phosphate
Specifications
Specifications
| Chemical Name or Material | (3RS)-N,N-Bis(1-methylethyl)-3-phenyl-3-(pyridin-2-yl)propan-1-amine Dihydrogen Phosphate |
| CAS | 2731375-73-6 |
| Analyte or Component Names | (3RS)-N,N-Bis(1-methylethyl)-3-phenyl-3-(pyridin-2-yl)propan-1-amine Dihydrogen Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H28 N2 . H3 O4 P |
| Certifications/Compliance | ISO 9001 |
| Synonym | Disopyramide Imp. B (EP), (3RS)-N,N-Bis(1-methylethyl)-3-phenyl-3-(pyridin-2-yl)propan-1-amine Dihydrogen Phosphate, Disopyramide Imp. B (EP) as Dihydrogen Phosphate, Disopyramide Impurity B as Dihydrogen Phosphate |
| InChI Formula | InChI=1S/C20H28N2.H3O4P/c1-16(2)22(17(3)4)15-13-19(18-10-6-5-7-11-18)20-12-8-9-14-21-20;1-5(2,3)4/h5-12,14,16-17,19H,13,15H2,1-4H3;(H3,1,2,3,4) |
| SMILES | CC(C)N(CCC(c1ccccc1)c2ccccn2)C(C)C.OP(=O)(O)O |
| IUPAC Name | 3-phenyl-N,N-di(propan-2-yl)-3-pyridin-2-ylpropan-1-amine;phosphoric acid |
| Show More |