Learn More
(3RS)-7-Chloro-5-(2-chlorophenyl)-2-oxo-2,3-dihydro-1H-1,4-benzodiazepin-3-ylAcetate (Lorazepam Acetate), Mikromol
Description
(3RS)-7-Chloro-5-(2-chlorophenyl)-2-oxo-2,3-dihydro-1H-1,4-benzodiazepin-3-ylAcetate (Lorazepam Acetate), Mikromol
Specifications
Specifications
| Chemical Name or Material | Lorazepam Acetate |
| CAS | 2848-96-6 |
| Analyte or Component Names | Lorazepam Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H12 Cl2 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | ((3RS)-7-Chloro-5-(2-chlorophenyl)-2-oxo-2,3-dihydro-1H-1,4-benzodiazepin-3-yl Acetate), Lorazepam Imp. B (EP), Lorazepam USP Related Compound A, Lorazepam USP RC A, (3RS)-7-Chloro-5-(2-chlorophenyl)-2-oxo-2,3-dihydro-1H-1,4-benzodiazepin-3-ylAcetate, Lorazepam Acetate, Lorazepam Related Compound A, Lorazepam Impurity B |
| InChI Formula | InChI=1S/C17H12Cl2N2O3/c1-9(22)24-17-16(23)20-14-7-6-10(18)8-12(14)15(21-17)11-4-2-3-5-13(11)19/h2-8,17H,1H3,(H,20,23) |
| Solution Type | Neat |
| SMILES | CC(=O)OC1N=C(c2ccccc2Cl)c3cc(Cl)ccc3NC1=O |
| Show More |