missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoylacetonitrile, 97%, Thermo Scientific™

Product Code. p-7022677
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483017 1 g 1g
11493017 5 g 5g
11403027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483017 Supplier Thermo Scientific Alfa Aesar Supplier No. A15564.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(Trifluoromethyl)benzoylacetonitrile is used as a pharmaceutical intermediate.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27328-86-5
Molecular Formula C10H6F3NO
Molecular Weight (g/mol) 213.159
MDL Number MFCD00052315
InChI Key GEPORLBYZLQDOB-UHFFFAOYSA-N
Synonym 3-trifluoromethylbenzoylacetonitrile, 3-oxo-3-3-trifluoromethyl phenyl propanenitrile, 3-trifluoromethyl benzoylacetonitrile, 2-cyano-3'-trifluoromethyl acetophenone, 3-oxo-3-3-trifluoromethyl-phenyl-propionitrile, pubchem12076, acmc-1cbhg, maybridge1_000123, 3-trifluoromethyl benzoyl acetonitrile
PubChem CID 520304
IUPAC Name 3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)CC#N

Specifications

Melting Point 58°C to 63°C
Quantity 1 g
Beilstein 2373428
Formula Weight 213.16
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzoylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.