accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFON

CAS RN 27328-86-5

IUPAC Name: 3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
Synonyms: 3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
Molecular Weight (g/mol): 213.16
Molecular Formula: C10H6F3NO
InChi Key: GEPORLBYZLQDOB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)CC#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)