Learn More
3-(quinolin-2-yl)propanoic Acid, TRC
Product Code. 30383930
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383930 25 mg -
30383125 50 mg 50mg
30383213 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30383930 Supplier Toronto Research Chemicals Supplier No. TRCE59244525MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39111-94-9
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.22
SMILES OC(=O)CCc1ccc2ccccc2n1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C12H11NO2/c14-12(15)8-7-10-6-5-9-3-1-2-4-11(9)13-10/h1-6H,7-8H2,(H,14,15)
Chemical Name or Material 3-(quinolin-2-yl)propanoic Acid