CAS RN 39111-94-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(quinolin-2-yl)propanoic Acid, TRC
CAS: 39111-94-9 Molecular Formula: C12H11NO2 Molecular Weight (g/mol): 201.22 SMILES: OC(=O)CCc1ccc2ccccc2n1