missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[f]quinoline, 97%, Thermo Scientific™

Product Code. 11491407
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491407 25 g 25g
11481407 5 g 5g
11401417 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491407 Supplier Thermo Scientific Alfa Aesar Supplier No. B24920.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methylbenzo[f]quinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-06-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00046906
InChI Key SUHRSZJZYUCLOD-UHFFFAOYSA-N
Synonym 3-methylbenzo f quinoline, benzo f quinoline, 3-methyl, beta-naphthoquinaldine, .beta.-naphthoquinaldine, unii-j67ocr12xn, j67ocr12xn
PubChem CID 6797
IUPAC Name 3-methylbenzo[f]quinoline
SMILES CC1=NC2=C(C=C1)C3=CC=CC=C3C=C2

Specifications

Melting Point 73°C to 78°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[f]quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.