accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3N

CAS RN 85-06-3

IUPAC Name: 3-methylbenzo[f]quinoline
Synonyms: 3-methylbenzo[f]quinoline
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: SUHRSZJZYUCLOD-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(C=CC3=CC=CC=C23)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results