Learn More
3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride, Mikromol™
Description
3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | 3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride |
| CAS | 2724726-76-3 |
| Analyte or Component Names | 3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C26 H27 N3 O6 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride, Nicardipine Hydrochloride Impurity A as Dihydrochloride, Nicardipine Impurity A as Dihydrochloride |
| InChI Formula | InChI=1S/C26H27N3O6.2ClH/c1-17-22(25(30)34-4)24(20-11-8-12-21(15-20)29(32)33)23(18(2)27-17)26(31)35-14-13-28(3)16-19-9-6-5-7-10-19;;/h5-12,15H,13-14,16H2,1-4H3;2*1H |
| SMILES | Cl.Cl.COC(=O)c1c(C)nc(C)c(C(=O)OCCN(C)Cc2ccccc2)c1c3cccc(c3)[N+](=O)[O-] |
| IUPAC Name | 3-O-[2-[benzyl(methyl)amino]ethyl] 5-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate;dihydrochloride |
| Show More |